Order Entry
Spain
ContactUsLinkComponent
Impurity and API standard, topiramate
Impurity and API standard, topiramate
  PROOMM1348.00
 :  
Impurity and API standard, topiramate
  PROOMM1348.00
 :  

 

  • Pk:
    100 mg
  • Standards:
    Reference standards
  • CAS No.:
    97240-79-4
  • Description:
    Topiramate
  • Formula:
    C₁₂H₂₁NO₈S
  • Description:
    Topiramate