Order Entry
Spain
ContactUsLinkComponent
Impurity and API standard, 3-Piperazin-1-yl-1,2-benzisothiazole hydrochloride
Impurity and API standard, 3-Piperazin-1-yl-1,2-benzisothiazole hydrochloride
Catalog # PROOMM1273.06
CAS Number:  
Impurity and API standard, 3-Piperazin-1-yl-1,2-benzisothiazole hydrochloride
Catalog # PROOMM1273.06
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Reference standards
  • CAS No.:
    87691-88-1
  • Description:
    3-Piperazin-1-yl-1,2-benzisothiazole hydrochloride
  • Formula:
    C₁₁H₁₄ClN₃S
  • Description:
    3-Piperazin-1-yl-1,2-benzisothiazole hydrochloride

Specifications

About this item