Impurity and API standard, diphemanil metilsulphate
PROOMM1163.00
:
- Pk:100 mg
- Standards:Reference standards
- CAS No.:62-97-5
- Description:Diphemanil metilsulphate
- Environmentally Preferable:
- Formula:C₂₁H₂₇NO₄S
- Description:Diphemanil metilsulphate