Order Entry
Spain
ContactUsLinkComponent
Impurity and API standard, 2-cyano-N-[4-(trifluoromethyl)phenyl]acetamide
Impurity and API standard, 2-cyano-N-[4-(trifluoromethyl)phenyl]acetamide
Catalog # PROOMM0859.08
CAS Number:  
Impurity and API standard, 2-cyano-N-[4-(trifluoromethyl)phenyl]acetamide
Catalog # PROOMM0859.08
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Reference standards
  • CAS No.:
    24522-30-3
  • Description:
    2-Cyano-N-[4-(trifluoromethyl)phenyl]acetamide
  • Formula:
    C₁₀H₇F₃N₂O
  • Description:
    2-Cyano-N-[4-(trifluoromethyl)phenyl]acetamide

Specifications

About this item