Impurity and API standard, 5-methylisoxazole-4-carboxylic Acid
Catalog # PROOMM0859.04
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:100 mg
- Standards:Reference standards
- CAS No.:42831-50-5
- Description:5-Methylisoxazole-4-carboxylic acid
- Environmentally Preferable:
- Formula:C₅H₅NO₃
- Description:5-Methylisoxazole-4-carboxylic acid