Order Entry
Spain
ContactUsLinkComponent
Impurity and API standard, 2,2'-binaphthalene-6,6'-dicarboxylic Acid
Impurity and API standard, 2,2'-binaphthalene-6,6'-dicarboxylic Acid
  PROOMM0681.03
 :  
Impurity and API standard, 2,2'-binaphthalene-6,6'-dicarboxylic Acid
  PROOMM0681.03
 :  

 

  • Pk:
    50 mg
  • Standards:
    Reference standards
  • CAS No.:
    932033-58-4
  • Description:
    2,2'-Binaphthalene-6,6'-dicarboxylic acid
  • Formula:
    C₂₂H₁₄O₄
  • Description:
    2,2'-Binaphthalene-6,6'-dicarboxylic acid