Order Entry
Spain
ContactUsLinkComponent
Impurity and API standard, 1-(2-methoxyphenyl)tricyclo[3.3.1.13,7]decane
Impurity and API standard, 1-(2-methoxyphenyl)tricyclo[3.3.1.13,7]decane
Catalog # PROOMM0681.01
CAS Number:  
Impurity and API standard, 1-(2-methoxyphenyl)tricyclo[3.3.1.13,7]decane
Catalog # PROOMM0681.01
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Reference standards
  • CAS No.:
    43109-77-9
  • Description:
    1-(2-Methoxyphenyl)tricyclo[3.3.1.13,7]decane
  • Formula:
    C₁₇H₂₂O
  • Description:
    1-(2-Methoxyphenyl)tricyclo[3.3.1.13,7]decane

Specifications

About this item