Impurity and API standard, fentanyl N-oxide hydrochloride
PROOMM0528.09
:
- Pk:100 mg
- Standards:Reference standards
- CAS No.:85893-37-4
- Description:N-Phenyl-N-[cis,trans-1-oxido-1-(2-phenylethyl)piperidin-4-yl]propanamide hydrochloride
- Formula:C₂₂H₂₈N₂O₂.ClH
- Description:N-Phenyl-N-[cis,trans-1-oxido-1-(2-phenylethyl)piperidin-4-yl]propanamide hydrochloride