Order Entry
Spain
ContactUsLinkComponent
Impurity and API standard, (2S)-1-[(1,1-dimethylethyl)amino]-3-[[4-[(2-hydroxyethyl)amino]-1,2,5-thiadiazol-3-yl]oxy]propan-2-ol
Impurity and API standard, (2S)-1-[(1,1-dimethylethyl)amino]-3-[[4-[(2-hydroxyethyl)amino]-1,2,5-thiadiazol-3-yl]oxy]propan-2-ol
Catalog # PROOMM0440.18
CAS Number:  
Impurity and API standard, (2S)-1-[(1,1-dimethylethyl)amino]-3-[[4-[(2-hydroxyethyl)amino]-1,2,5-thiadiazol-3-yl]oxy]propan-2-ol
Catalog # PROOMM0440.18
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Reference standards
  • CAS No.:
    75014-26-5
  • Description:
    (2S)-1-[(1,1-dimethylethyl)amino]-3-[[4-[(2-hydroxyethyl)amino]-1,2,5-thiadiazol-3-yl]oxy]propan-2-ol
  • Formula:
    C₁₁H₂₂N₄O₃S
  • Description:
    (2S)-1-[(1,1-Dimethylethyl)amino]-3-[[4-[(2-hydroxyethyl)amino]-1,2,5-thiadiazol-3-yl]oxy]propan-2-ol

Specifications

About this item