Impurity and API standards, 1-[2-[(2RS)-2,3-dihydroxypropoxy]phenyl]-3-phenylpropan-1-one
Catalog # PROOMM0405.05
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:50 mg
- Standards:Reference standards
- CAS No.:91401-73-9
- Description:1-[2-[(2RS)-2,3-dihydroxypropoxy]phenyl]-3-phenylpropan-1-one
- Formula:C₁₈H₂₀O₄
- Description:1-[2-[(2RS)-2,3-Dihydroxypropoxy]phenyl]-3-phenylpropan-1-one