Order Entry
Spain
ContactUsLinkComponent
Impurity and API standards, 3-Ethyl 5-Methyl 2-[(2-Aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methylpyridine-3,5-dicarboxylate fumarate
Impurity and API standards, 3-Ethyl 5-Methyl 2-[(2-Aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methylpyridine-3,5-dicarboxylate fumarate
Certificates
Impurity and API standards, 3-Ethyl 5-Methyl 2-[(2-Aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methylpyridine-3,5-dicarboxylate fumarate

Specifications

  • CAS No.:
    113994-41-5
  • Formula:
    C₂₀H₂₃ClN₂O₅
  • Description:
    3-ethyl 5-methyl 2-[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methylpyridine-3,5-dicarboxylate fumarate

Specifications

Frequently Bought Together