Order Entry
Spain
ContactUsLinkComponent
Impurity and API standards, ethoxycetirizine dihydrochloride
Impurity and API standards, ethoxycetirizine dihydrochloride
Catalog # PROOMM0380.05
CAS Number:  
Impurity and API standards, ethoxycetirizine dihydrochloride
Catalog # PROOMM0380.05
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Reference standards
  • CAS No.:
    83881-56-5
  • Description:
    (RS)-2-[2-[2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]ethoxy]acetic acid dihydrochloride (ethoxycetirizine dihydrochloride)
  • Formula:
    C₂₃H₂₉ClN₂O₄·₂ClH
  • Description:
    (RS)-2-[2-[2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]ethoxy]acetic acid Dihydrochloride (Ethoxycetirizine Dihydrochloride)

Specifications

About this item