Order Entry
Spain
ContactUsLinkComponent
Impurity and API standard, 5-Methyl-5-phenylpyrimidine-2,4,6(1H,3H,5H)-trione
Impurity and API standard, 5-Methyl-5-phenylpyrimidine-2,4,6(1H,3H,5H)-trione
Catalog # PROOMM0265.08
CAS Number:  
Impurity and API standard, 5-Methyl-5-phenylpyrimidine-2,4,6(1H,3H,5H)-trione
Catalog # PROOMM0265.08
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Reference standards
  • CAS No.:
    76-94-8
  • Description:
    5-Methyl-5-phenylpyrimidine-2,4,6(1H,3H,5H)-trione
  • Formula:
    C₁₁H₁₀N₂O₃
  • Description:
    5-Methyl-5-phenylpyrimidine-2,4,6(1H,3H,5H)-trione

Specifications

About this item