Order Entry
Spain
ContactUsLinkComponent
Organic reference standards, Potassium sucrose octasulphate, Ph. Eur., RS
Organic reference standards, Potassium sucrose octasulphate, Ph. Eur., RS
Catalog # EDQMY0001275
Supplier:  EDQM
CAS Number:  
Organic reference standards, Potassium sucrose octasulphate, Ph. Eur., RS
Catalog # EDQMY0001275
Supplier:  EDQM
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Reference standard
  • CAS No.:
    73264-44-5
  • Description:
    Potassium sucrose octasulphate
  • Formula:
    C₁₂H₁₄K₈O₃₅S₈
  • Description:
    Potassium sucrose octasulfate

Specifications

About this item