Order Entry
Spain
ContactUsLinkComponent
Impurity and API standard, 2,4-bis[(furan-2-ylmethyl)amino]-5-sulphamoylbenzoic acid
Impurity and API standard, 2,4-bis[(furan-2-ylmethyl)amino]-5-sulphamoylbenzoic acid
  PROOMM0014.04
 :  
Impurity and API standard, 2,4-bis[(furan-2-ylmethyl)amino]-5-sulphamoylbenzoic acid
  PROOMM0014.04
 :  

 

  • Pk:
    100 mg
  • Standards:
    Reference standards
  • CAS No.:
    5046-19-5
  • Description:
    2,4-bis[(furan-2-ylmethyl)amino]-5-sulphamoylbenzoic acid
  • Formula:
    C₁₇H₁₇N₃O₆S
  • Description:
    2,4-Bis[(furan-2-ylmethyl)amino]-5-sulphamoylbenzoic Acid