Order Entry
Spain
ContactUsLinkComponent
Powder pillows, potassium persulphate
Powder pillows, potassium persulphate
Catalog # HACH2084766
Supplier:  Hach
CAS Number:  
undefined
Powder pillows, potassium persulphate
Catalog # HACH2084766
Supplier:  Hach
CAS Number:  
Specifications
  • Chemical method:
    Persulfate UV oxidation
  • Pk:
    50
  • Description:
    Powder pillows, potassium persulphate
Specifications
About this item
Powder reagent in sealed package for phosphonate test and total phosphate TNT.