Order Entry
Spain
ContactUsLinkComponent
 
D-(+)-Glucose, anhydrous, granular, AR® ACS, Macron Fine Chemicals™
 
undefined
D-(+)-Glucose, anhydrous, granular, AR® ACS, Macron Fine Chemicals™
 

 

  • Meets ACS Reagent Chemical Requirements:
    ≤0.01%
  • For Laboratory,Research,or Manufacturing Use:
    ≤5 ppm
  • ACS - Chloride (Cl):
    ≤0.01 %
  • ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - Iron (Fe):
    ≤5 ppm
  • ACS - Loss on Drying at 105°C:
    ≤0.2 %
  • ACS - Residue after Ignition:
    ≤0.02 %
  • ACS - Starch:
    Passes Test
  • ACS - Sulfate and Sulfite (as SO₄):
    ≤0.005 %
  • ACS - Specific Rotation [Å]²⁵^D (+):
    52.5 - 53.0 Degree
  • ACS - Titrable Acid (meq/g):
    ≤0.002

 

Frequently Bought Together