
Amonio acetato ≥97%, en cristales, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:631-61-8
:
Formula:H₃CCOONH₄
MDL Number:MFCD00013066
:77.08 g/mol
Merck Index:13,00495
Melting Pt:114 °C
Density:1,17 g/cm³ (20 °C)
Flash Pt:136 °C
EINECS:211-162-9
:GHS07
Storage Temperature:Ambiente
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:≥97%
- Assay (CH₃COONH₄) (by acid-base titration):≥97 %
- Insoluble Matter:≤0.005 %
- Nitrate (NO₃):≤0.001 %
- pH of 5% Solution at 25°C:6.7 - 7.3
- Residue after Ignition:≤0.010 %
- Solution (20:100) - clear, colorless, complete:Passes Test
- Sulfate (SO₄):≤0.001 %
- Water (H₂O):≤2.5 %
- Chloride (Cl):≤5 ppm
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
- Trace Impurities - Iron (Fe):≤5 ppm
Specification Test Results
Frequently Bought Together