Order Entry
Spain
ContactUsLinkComponent
Acetic acid glacial 99.5-100.5% USP, FCC, ACS, Macron Fine Chemicals™
Acetic acid glacial 99.5-100.5% USP, FCC, ACS, Macron Fine Chemicals™
Supplier:  Avantor
Certificates
undefined
Acetic acid glacial 99.5-100.5% USP, FCC, ACS, Macron Fine Chemicals™
Supplier:  Avantor
Certificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specification Test Results

  • Meets U.S.P Requirements:
  • Meets ACS Reagent Chemical Requirements:
  • Meets F.C.C. Requirements:
  • GMP Manufactured Product:
  • Food GMP Manufactured Product:
  • IMPORTANT: Material will freeze if stored below 17 °C (63°F).:
  • CAUTION: For Manufacturing, processing or repackaging:
  • Bulk Food Chemical:
  • Bulk Pharmaceutical Chemical:
  • USP - Assay (CH₃COOH):
    99.5 - 100.5 %
  • USP - Chloride (Cl) (USP):
    Passes Test
  • USP - Congealing Temperature:
    ≥15.6 °C
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Readily Oxidizable Substances:
    Passes Test
  • USP - Sulfate (USP)(SO₄):
    Passes Test
  • USP - Limit of Nonvolatile Residue:
    ≤0.005 %
  • FCC - Assay (CH₃COOH):
    99.7 - 100.5 %
  • FCC - Identification A:
    Passes Test
  • FCC - Identification B:
    Passes Test
  • FCC - Lead (Pb):
    ≤0.5 mg/kg
  • FCC - Limit of Nonvolatile Residue:
    ≤0.005 %
  • FCC - Readily Oxidizable Substances:
    Passes Test
  • FCC - Solidification Point:
    ≥15.6 °C
  • ACS - Assay (by GC, corrected for water):
    ≥99.7 %
  • ACS - Acetic Anhydride ((CH₃CO)₂O):
    ≤0.01 %
  • ACS - Chloride (Cl):
    ≤1 ppm
  • ACS - Color (APHA):
    ≤10
  • Dilution Test:
    Passes Test
  • Trace Impurities - ACS - Iron (Fe):
    ≤0.200 ppm
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤0.5 ppm
  • ACS - Substances Reducing Dichromate:
    Passes Test
  • ACS - Substances Reducing Permanganate:
    Passes Test
  • ACS - Sulfate (SO₄):
    ≤1 ppm
  • ACS - Titrable Base (meq/g):
    ≤0.0004
  • Copper (Cu):
    ≤0.00001 %
  • Nickel (Ni):
    ≤0.00001 %
  • Residue after Evaporation:
    ≤0.001 %
  • Appearance (clear, colorless liquid):

Specification Test Results

Related Information

Frequently Bought Together