Acetic acid 36.0 - 37.0% NF, Macron Fine Chemicals™
Catalog # 2488-44
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Bulk Pharmaceutical Chemical:36.0 - 37.0 %
- NF - Assay (CH₃COOH):36.0 - 37.0 %
- NF - Chloride (Cl):Passes Test
- NF - Identification:Passes Test
- NF - Nonvolatile Residue:≤0.005 %
- NF - Readily Oxidizable Substances:Passes Test
- NF - Sulfate (SO₄):Passes Test
Specification Test Results
About this item
:
:60.05 g/mol
UN:2790
ADR:8,III
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152