Order Entry
Spain
ContactUsLinkComponent
 
Size exclusion chromatography columns, Superdex™ 30 Increase
 :  Cytiva
 
undefined
Size exclusion chromatography columns, Superdex™ 30 Increase
 :  Cytiva
 

 

  • Catalog No:
  • 29-2197-58
  • 29-2197-57
  • Max. pressure:
  • 30 bar
  • 30 bar
  • pH range:
  • 3 - 12
  • 3 - 12
  • Storage:
  • 4 to 30 °C, 20% ethanol
  • 4 to 30 °C, 20% ethanol

 

Frequently Bought Together