Order Entry
Spain
ContactUsLinkComponent
Multi-channel pipettes, mechanical, variable volume, Eppendorf Research® plus (IVD marked)
Multi-channel pipettes, mechanical, variable volume, Eppendorf Research® plus (IVD marked)
Catalog # 613-0883
Supplier:  EPPENDORF
undefined
Multi-channel pipettes, mechanical, variable volume, Eppendorf Research® plus (IVD marked)
Catalog # 613-0883
Supplier:  EPPENDORF

Specifications

  • Colour code:
    Grey
  • Volume:
    0,5 - 10 µl
  • Seq:
    01
  • Environmentally Preferable:
  • Increment:
    0,01 µl
  • Accuracy:
    0,5 µl: ±12,0 % / ±0,06 µl
    1 µl: ±8,0 % / ±0,08 µl
    5 µl: ±4,0 % / ±0,2 µl
    10 µl: ±2,0 % / ±0,2 µl
  • Imprecision:
    0,5 µl: ±8,0 % / ±0,04 µl
    1 µl: ±5,0 % / ±0,05 µl
    5 µl: ±2,0 % / ±0,1 µl
    10 µl: ±1,0 % / ±0,1 µl
  • Pk:
    1
  • Recommended tips:
    epT.I.P.S.® 20 µl

Specifications

About this item

Versatile, ergonomic pipette with innovative design, meets the highest needs in precision and accuracy.

  • Extremely lightweight
  • Innovative re-designed principle of piston-stroke pipetting, piston made of organic polymer Fortron® with great chemical, thermal and mechanical resistance
  • Lowest aspirating, dispensing, blow-out and tip ejection forces
  • Spring-loaded, retractable nose cone to ensure tight seal and prolong nose cone life, as well as exact tip alignment
  • Individual channel removal for quick and easy servicing
  • Fully autoclavable

Designed and constructed for clinical applications (clinical human diagnostics). Ideal for low-contamination transfer of liquids, in particular for samples from the human body and for reagents within the scope of an in vitro diagnostic application in order to allow the in vitro diagnostic medical device to be used as intended. This pipette is an in vitro diagnostic device according to directive 98/79/EC of the european parliament and the council dated october 27, 1998. It is intended exclusively for indoor use and for operation by qualified staff.

Certifications: With CE marking.