Specifications
- Antibody type:Primary
- Antigen name:Carbonic Anhydrase 11
- Antigen symbol:CA11
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:770
- Antigen synonyms:CARPX1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Carbonic anhydrases (CAs) are members of a large family of zinc metalloenzymes responsible for catalyzing the reversible hydration of carbon dioxide. CAs show extensive diversity in their distribution and subcellular localization. They are involved in a variety of biological processes, including calcification, bone resorption, respiration, acid-base balance and the formation of aqueous humor, saliva, gastric juice and cerebrospinal fluid. CA XI, also referred to as carbonic anhydrase-related protein 11 precursor (CA-RP XI) or carbonic anhydrase-related protein 2 (CA-RP II), is a member of the carbonic anhydrase family that lacks two of the three Zn-binding motifs essential for carbonic anhydrase activity. For this reason, CA XI does not exhibit catalytic activity. It is expressed primarily in brain but is also found in spinal cord and thyroid. CA XI may play a role in brain development.Tissue specificity:Expressed abundantly in the brain with moderate expression also present in spinal cord and thyroid.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CA11
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat