Order Entry
Spain
ContactUsLinkComponent
Anti-RUNX3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anti-RUNX3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-4848R-A350
Supplier:  Bioss
Anti-RUNX3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-4848R-A350
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Runt-related Transcription Factor 3
  • Antigen symbol:
    RUNX3
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 350
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    864
  • Antigen synonyms:
    AML2|Core-binding factor subunit alpha-3|RUNX3|SL3-3 enhancer factor 1 alpha C subunit|PEBP2A3|Runt-related transcription factor 3|Oncogene AML-2|CBFA3|Polyomavirus enhancer-binding protein 2 alpha C subunit|Acute myeloid leukemia 2 protein|SL3/
  • Modification:
    phospho-Ser338
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

RUNX3 binds to the core site of murine Leukemia virus, the core sequences in the enhancer of the polyomavirus, and also to the enhancers of the T-cell receptor genes. May be involved in the control of cellular proliferation and/or differentiation (By similarity). Heterodimer of an alpha and a beta subunit. The alpha subunit binds DNA as a monomer and through the Runt domain.DNA-binding is increased by heterodimerization. Interacts with TLE1 and SUV39H1, Subcellular location in Nucleus.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RUNX3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human