Anti-CR2 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-3792R-CY3
Proveedor: Bioss
Especificaciones
- Antibody type:Primary
- Antigen name:Complement Component (3d/Epstein Barr virus) receptor 2
- Antigen symbol:CR2
- Clonality:Polyclonal
- Conjugation:Cy3®
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:1380
- Antigen synonyms:SLEB9|CR|CVID7|CR2|CD21|C3DR
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:1050-1092/1092
- Purification:Purified by Protein A
- Pk:100 µl
Especificaciones
Acerca de este artículo
Receptor for complement C3Dd, for the Epstein-Barr virus on human B-cells and T-cells and for HNRPU. Participates in B lymphocytes activation.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: CR2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity: