Anti-NKX2-5 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
BOSSBS-2054R-A647
: Bioss
- Antibody type:Primary
- Antigen name:NK2 homeobox 5
- Antigen symbol:NKX2-5
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:18091
- Antigen synonyms:VSD3|NKX4-1|NKX2.5|CHNG5|CSX1|NKX2E|CSX|HLHS2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:80-130/318
- Purification:Purified by Protein A
- Pk:100 µl
Implicated in commitment to and/or differentiation of the myocardial lineage. Acts as a transcriptional activator of ANF in cooperation with GATA4. It is transcriptionally controlled by PBX1 and acts as a transcriptional repressor of CDKN2B. Together with PBX1, it is required for spleen development through a mechanism that involves CDKN2B repression.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Nkx2.5/Cardiac-specic homeobox 1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat