Anti-TEP1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-1908R-FITC
Supplier: Bioss
Specifications
- Tipo de anticuerpo:Primario
- Nombre del antígeno:Telomerase-associated Protein 1
- Simbolo antígeno:TEP1
- Clonalidad:Polyclonal
- Conjugado:FITC (Fluorescein Isothiocyanate)
- Huésped:Rabbit
- InmunoQuímica:Yes
- Isotipo:IgG
- Reactividad:Human,Rat,Mouse
- Adsorción cruzada:No
- Forma:liquid
- ID del gen:7011
- Sinónimos de antígenos:TLP1|VAULT2|TP1|Telomerase protein 1|Telomerase-associated protein 1|TROVE1|p240|Telomerase protein component 1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Temperatura de almacenamiento:Store at 4°C for 12 months
- Concentración:1 μg/μl
- Shipping temperature:4°C
- Inmunógeno:1740-1780/2627
- Purification:Purified by Protein A
- Env:100 µl
Specifications
About this item
Component of the telomerase ribonucleoprotein complex that is essential for the replication of chromosome termini. Also component of the ribonucleoprotein vaults particle, a multi-subunit structure involved in nucleo-cytoplasmic transport. Responsible for the localizing and stabilizing vault RNA (vRNA) association in the vault ribonucleoprotein particle. Binds to TERC (By similarity).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TEP1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat