Anti-GABAA Rgamma1 Rabbit Polyclonal Antibody [clone: K51]
BWRLBS3301
- Antibody type:Primary
- Antigen symbol:GABAA R gamma1
- Clonality:Polyclonal
- Clone:K51
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 20-72 of Human GABAA Rgamma1.
GABAA R?1 (K51) pAb detects endogenous levels of GABAA R?1 protein.
Type: Primary
Antigen: GABAA Rgamma1
Clonality: Polyclonal
Clone: K51
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat