Anti-DGK-epsilon Rabbit Polyclonal Antibody [clone: S194]
Catalog # BWRLBS2281
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:DGK- epsilon
- Clonality:Polyclonal
- Clone:S194
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 180-230 of Human DGK-epsilon.
DGK-? (S194) pAb detects endogenous levels of DGK-? protein.
Type: Primary
Antigen: DGK-epsilon
Clonality: Polyclonal
Clone: S194
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat