Anti-VEGF-D Rabbit Polyclonal Antibody [clone: I202]
Catalog # BWRLBS3675
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:VEGF-D
- Clonality:Polyclonal
- Clone:I202
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 171-220 of Human VEGF-D.
VEGF-D (I202) pAb detects endogenous levels of VEGF-D protein.
Type: Primary
Antigen: VEGF-D
Clonality: Polyclonal
Clone: I202
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat