Anti-ZNF75 Rabbit Polyclonal Antibody [clone: K107]
Catalog # BWRLBS2010
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:ZNF75
- Clonality:Polyclonal
- Clone:K107
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 80-120 of Human ZNF75.
ZNF75 (K107) pAb detects endogenous levels of ZNF75 protein.
Type: Primary
Antigen: ZNF75
Clonality: Polyclonal
Clone: K107
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human