Order Entry
Spain
Orders LinkContactUsLinkComponent
Ascorbic acid titrant solution
Ascorbic acid titrant solution
Dropping bottle, marked, 100 mL
Catalog # HACH2308232
Irritant
Supplier:   Hach
Ascorbic acid titrant solution
Catalog # HACH2308232
Supplier:  Hach
CAS Number:  
Ascorbic acid titrant solution
Ascorbic acid titrant solution
Dropping bottle, marked, 100 mL
CAS Number:  
Price
33,90 €
Log in for availability
1 * 100 mL
Product Details & Documents
This titrant is used for the determination of ascorbic acid in juice based beverages, by drop-count titration.
Specifications
Specifications
  • Pack type:
    Dropping bottle, marked
  • Pk:
    100 mL
Specifications