Anti-DSG2 Mouse Monoclonal Antibody [clone: 8 E5]
Catalog # ABNOMAB14503
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Desmoglein 2 Recombinant
- Antigen symbol:DSG2
- Clonality:Monoclonal
- Clone:8 E5
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:1829
- Storage buffer:In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
- Storage temperature:Store at 4° C.
- Immunogen:Recombinant protein corresponding to full length human DSG2.
- Purification:Protein A/G purification
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant human DSG2.
Type: Primary
Antigen: DSG2
Clonality: Monoclonal
Clone: 8 E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human