Order Entry
Spain
ContactUsLinkComponent
Protein gel stain, GelCode™ SilverSNAP® silver stain II, Pierce™
Protein gel stain, GelCode™ SilverSNAP® silver stain II, Pierce™
  PIER24612
 :  
undefined
Protein gel stain, GelCode™ SilverSNAP® silver stain II, Pierce™
  PIER24612
 :  
  • Pk:
    1 L
  • Description:
    Protein gel stain, GelCode™ SilverSNAP® Silver Stain II, Pierce™
GelCode™ SilverSNAP® Silver Stain II is a fast, flexible stain that delivers consistently reliable staining performance.
  • Low and uniform background
  • Detection as low as 0,25 ng of protein
  • Staining can be completed in under 50 minutes
  • Compatible with a wide assortment of homemade and precast gels
  • Flexible protocol without altering sensitivity or background
Suitable for use with 1- or 2-D PAGE and IEF gels and used to stain DNA or RNA following electrophoresis.