Inlet liners
Supplier: Restek
Jump to:
About this item
These liners can be used with Agilent GCs.
Ø int. = Nominal diameter at syringe needle expulsion point
29 Options Available Below
Product Details & Documents
These liners can be used with Agilent GCs.
Ø int. = Nominal diameter at syringe needle expulsion point
Product Family Options
Liners for Agilent GCs
Product Information
- Ext.ØØ int.LengthAvailabilityPrice
- 568-2333Split, Siltek, with wool, universal use with Agilent 7673 autosamplerSplit, Siltek, with wool, universal use with Agilent 7673 autosampler6,3 mm4,0 mm78,5 mmDiscontinued-
- REST20782-213Split, Siltek, with wool, universal use with Agilent 7673 autosamplerSplit, Siltek, with wool, universal use with Agilent 7673 autosampler6,3 mm4,0 mm78,5 mmLog in for availability1 * 5 UN262,00 €
- 568-2334Split, with wool, universal use with Agilent 7673 autosamplerSplit, with wool, universal use with Agilent 7673 autosampler6,3 mm4,078,5Log in for availability1 * 5 UN105,00 €
- REST20773Splitless, for trace samples >2 µl
- REST22406Splitless, gooseneck, with wool, for trace samples >2 µlSplitless, gooseneck, with wool, for trace samples >2 µl6,5 mm4,0 mm78,5 mmLog in for availability1 * 5 UN152,00 €
- 568-2327Splitless, Siltek, for trace samples >2 µlSplitless, Siltek, for trace samples >2 µl6,5 mm4,0 mm78,5 mmDiscontinued-
- REST20773-214Splitless, Siltek, for trace samples >2 µl
- REST20707Cyclosplitter for dirty samples, many injections before cleaning requiredCyclosplitter for dirty samples, many injections before cleaning required6,3 mm4,0 mm78,5 mmLog in for availability1 * 5 UN266,00 €
- REST20710Cup splitter for high and low MW compoundsCup splitter for high and low MW compounds6,3 mm4,0 mm78,5 mmLog in for availability1 * 5 UN247,00 €
- REST20771Drilled uniliner for trace, active samples, high recovery and linearityDrilled uniliner for trace, active samples, high recovery and linearity6,3 mm4,0 mm78,5 mmLog in for availability1 * 5 UN474,00 €
- REST20796Single taper splitless, for trace samples >2 µlSingle taper splitless, for trace samples >2 µl6,5 mm2,0 mm78,5 mmLog in for availability1 * 5 UN164,00 €
- REST20799Single taper splitless, for trace samples >2 µlSingle taper splitless, for trace samples >2 µl6,5 mm4,0 mm78,5Log in for availability1 * 5 UN138,00 €
- REST20785Double taper splitless, for trace and active samples >2 µlDouble taper splitless, for trace and active samples >2 µl6,5 mm4,0 mm78,5 mmLog in for availability1 * 5 UN195,00 €
- REST20896Cyclo double taper, for trace, active and dirty samples >2 µlCyclo double taper, for trace, active and dirty samples >2 µl6,5 mm4,0 mm78,5 mmLog in for availability1 * 5 UN539,00 €
- REST20908Cyclo double taper, for trace, active and dirty samples >2 µlCyclo double taper, for trace, active and dirty samples >2 µl6,5 mm2,0 mm78,5 mmLog in for availability1 * 5 UN399,00 €
- REST20913Splitless (quartz), for trace samples <2 µlSplitless (quartz), for trace samples <2 µl6,5 mm4,0 mm78,5 mmLog in for availability1 * 5 UN192,00 €
- REST20915Splitless (quartz), for trace samples <2 µlSplitless (quartz), for trace samples <2 µl6,5 mm2,0 mm78,5 mmLog in for availability1 * 5 UN204,00 €
- REST21055Drilled uniliner for trace, active samples, high recovery and linearityDrilled uniliner for trace, active samples, high recovery and linearity6,3 mm4,0 mm78,5 mmLog in for availability1 * 5 UN433,00 €
Liners for Varian GCs
Product Information
- Ext.ØØ int.LengthAvailabilityPrice
- REST21079Spilt, with wool, universal
- REST20898Splitless, cyclo double gooseneck, for trace, dirty, active samples up to 4 µl, for Varian 1075/1077Splitless, cyclo double gooseneck, for trace, dirty, active samples up to 4 µl, for Varian 1075/10776,3 mm4,0 mm74 mmDiscontinued-
- 568-0275Split, with wool, universal use with rapid autosamplers, for Varian 1075/1077Split, with wool, universal use with rapid autosamplers, for Varian 1075/10776,3 mm4,0 mm72 mmLog in for availability1 * 5 UN224,00 €
- REST20848Splitless, double gooseneck, for trace, active samples up to 4 µl, for Varian 1075/1077Splitless, double gooseneck, for trace, active samples up to 4 µl, for Varian 1075/10776,3 mm4,0 mm74Discontinued-
- REST20849Splitless, double gooseneck, for trace, active samples up to 4 µl, for Varian 1075/1077Splitless, double gooseneck, for trace, active samples up to 4 µl, for Varian 1075/10776,34,0 mm74 mmDiscontinued-
- 568-2357Split, with glass frit, universal for Varian 1177Split, with glass frit, universal for Varian 11776,3 mm4,0 mm78,5 mmLog in for availability1 * 5 UN268,00 €
Liners for Shimadzu GCs
Product Information
- Ext.ØØ int.LengthAvailabilityPrice
- 568-2351Split/splitless, Siltek, with wool, universal for most common analysesSplit/splitless, Siltek, with wool, universal for most common analyses5,0 mm3,5 mm95 mmDiscontinued-
- REST20956-213Split/splitless, Siltek, with wool, universal for most common analysesSplit/splitless, Siltek, with wool, universal for most common analyses5,0 mm3,5 mm95 mmLog in for availability1 * 5 UN308,00 €
- REST20956Split/splitless, with wool, universal for most common analysesSplit/splitless, with wool, universal for most common analyses5,0 mm3,5 mm95 mmLog in for availability1 * 5 UN173,00 €
- REST20861Split, universal for most common analyses
Liners for Perkin-Eklemer GCs
Product Information
- Ext.ØØ int.LengthAvailabilityPrice
- REST20732Splitless, for trace samplesSplitless, for trace samples5,0 mm2,0 mm100 mmDiscontinued-
- REST20740Cup splitter, high & low MW compoundsCup splitter, high & low MW compounds5,0 mm3,5 mm100 mmDiscontinued-
- REST20838Auto SYS open top uniliner with deactivated wool for trace, dirty, active samples, high recovery and linearityAuto SYS open top uniliner with deactivated wool for trace, dirty, active samples, high recovery and linearity6,2 mm4,0 mm92,1 mmLog in for availability1 * 5 UN351,00 €
- REST20840Auto SYS cyclo uniliner for trace, dirty, high MW active samples, high linearityAuto SYS cyclo uniliner for trace, dirty, high MW active samples, high linearity6,2 mm4,0 mm92,1 mmLog in for availability1 * 5 UN435,00 €
- REST21273Auto SYS splitless, headspace & purge & trapAuto SYS splitless, headspace & purge & trap6,2 mm1,0 mm92,1 mmLog in for availability1 * 5 UN229,00 €
- REST21293Auto SYS drilled uniliner (hole near bottom) for trace, active samples, high recovery and linearityAuto SYS drilled uniliner (hole near bottom) for trace, active samples, high recovery and linearity6,2 mm4,0 mm92,1 mmDiscontinued-
Liners for Thermo Scientific GCs
Product Information
- Ext.ØØ int.LengthAvailabilityPrice
- 568-2345Splitless, Siltek, for trace samples, for Trace™, 8000, 8000 TOP & Focus SSLSplitless, Siltek, for trace samples, for Trace™, 8000, 8000 TOP & Focus SSL8,0 mm3,0 mm105 mmDiscontinued-
- REST20943-214Splitless, Siltek, for trace samples, for Trace™, 8000, 8000 TOP & Focus SSLSplitless, Siltek, for trace samples, for Trace™, 8000, 8000 TOP & Focus SSL8,0 mm3,0 mm105 mmLog in for availability1 * 5 UN338,00 €
- REST20943Splitless, for trace samples, for Trace™, 8000, 8000 TOP & Focus SSLSplitless, for trace samples, for Trace™, 8000, 8000 TOP & Focus SSL8,0 mm3,0 mm105 mmLog in for availability1 * 5 UN249,00 €
- REST20946Splitless, for trace samples, for Trace™, 8000, 8000 TOP & Focus SSLSplitless, for trace samples, for Trace™, 8000, 8000 TOP & Focus SSL8,0 mm5,0 mm105 mmLog in for availability1 * 5 UN241,00 €
- REST21029Splitless precision liner with deactivated wool for dirty samples, trace samplesSplitless precision liner with deactivated wool for dirty samples, trace samples8,0 mm5,0 mm105 mmLog in for availability1 * 5 UN273,00 €