Anti-ARFGAP1 Rabbit Polyclonal Antibody
Catalog # BSBTA04959
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:ARFGAP1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for ADP-ribosylation factor GTPase-activating protein 1(ARFGAP1) detection. Tested with WB in Human, Mouse, Rat.
Type: Primary
Antigen: ARFGAP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat