Anti-AKT1 Rabbit Polyclonal Antibody
BSBTA00024
- Antibody type:Primary
- Antigen symbol:AKT1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for RAC-alpha serine/threonine-protein kinase(Akt1) detection. Tested with WB in Mouse, Rat.
Type: Primary
Antigen: AKT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat