Order Entry
Spain
ContactUsLinkComponent
Polystyrene Test Film - MIR
Polystyrene Test Film - MIR
Catalog # PERCL1202057
Supplier:  PerkinElmer
CAS Number:  
Polystyrene Test Film - MIR
Catalog # PERCL1202057
Supplier:  PerkinElmer
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Polystyrene MIR reference card
Specifications
About this item
This test film is an easy-to-use standard to check frequency, accuracy and any changes.
This reference material mounts easily in a FT-IR spectrometer, allowing the user to quickly and easily check the mid infrared frequencies and overall accuracy of the FT-IR spectrometer.
The Polystyrene Test Film is 0.038 mm thickness and untraceable.