Order Entry
Spain
ContactUsLinkComponent
Calbiochem® Human recombinant receptor-associated Protein (RAP) (from E. coli)
Calbiochem® Human recombinant receptor-associated Protein (RAP) (from E. coli)
Catalog # 553506-50
CAS Number:  
Calbiochem® Human recombinant receptor-associated Protein (RAP) (from E. coli)
Catalog # 553506-50
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Protein/peptide name:
    Receptor-associated Protein (RAP)

Specifications

About this item

Recombinant humman Receptor-associated Protein (RAP) fused to GST, expressed in E. coli and subsequently cleaved with thrombin to remove GST. Antagonizes the ligand binding of certain members of the low-density lipoprotein receptor family such as low density lipoprotein receptor-related protein (LRP) and very low-density lipoprotein (VLDL) receptor. Promotes the proper folding and subsequent exocytic trafficking of the wild-type low density lipoprotein receptor (LDLR) and several of its class 2 mutants. Expressed as a GST-fusion protein that is subsequently cleaved with thrombin. Typical concentrations to block LRP or VLDL receptor are 200-300 nM. For antagonism of LDLR, 1 µM is recommended.