Nut driver
Nut driver, 1/4"
Catalog # REST21076
Supplier: Restek
Jump to:
Product Details & Documents
This nut driver is ideal for changing Agilent 5890/6890/6850 FID jets and securely grips jet for easy removal or installation.
Specifications
- Description:Nut driver, 1/4"