Order Entry
Spain
ContactUsLinkComponent
Anti-DV1/DV2/DV3/DV4 Mouse Monoclonal Antibody [clone: D1-11]
Anti-DV1/DV2/DV3/DV4 Mouse Monoclonal Antibody [clone: D1-11]
  ABNOMAB12427
 :  Abnova
Anti-DV1/DV2/DV3/DV4 Mouse Monoclonal Antibody [clone: D1-11]
  ABNOMAB12427
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Dengue virus 1, 2, 3 and 4
  • Antigen symbol:
    DV1/DV2/DV3/DV4
  • Clonality:
    Monoclonal
  • Clone:
    D1-11
  • Host:
    Mouse
  • Isotype:
    IgG
  • Reactivity:
    Virus
  • Western blot:
    Yes
  • Form:
    Liquid
  • Storage buffer:
    In PBS, pH 7.2 (0.09% sodium azide)
  • Immunogen:
    A mixture of dengue virus 1, 2, 3 and 4.
  • Purification:
    Antigen Affinity Purification
  • Size:
    1 MG
  • Pk:
    1 mg

 

 

Mouse monoclonal antibody raised against virus Dengue Envelope Protein.

Type: Primary
Antigen: DV1/DV2/DV3/DV4
Clonality: Monoclonal
Clone: D1-11
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Viruses