Anti-Envelope Protein Mouse Monoclonal Antibody [clone: M86648]
ABNOMAB14217
: Abnova
- Antibody type:Primary
- Antigen name:Envelope Protein
- Clonality:Monoclonal
- Clone:M86648
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Virus
- Form:Liquid
- Storage buffer:In 0.015 M potassium phosphate, 0.85% NaCl, pH 7.2 (0.05% sodium azide).
- Immunogen:Recombinant protein corresponding to amino acids 291-690 of Zika virus envelope protein.
- Purification:DEAE column chromatography
- Size:500 µG
- Pk:500 µG
Mouse monoclonal antibody raised against partial recombinant Zika virus envelope protein.
Type: Primary
Antigen: Envelope Protein
Clonality: Monoclonal
Clone: M86648
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Viruses