Anti-NS1 Mouse Monoclonal Antibody [clone: M86641]
Catalog # ABNOMAB14219
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:NS1
- Clonality:Monoclonal
- Clone:M86641
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Virus
- Form:Liquid
- Storage buffer:In 0.015 M potassium phosphate, 0.85% NaCl, pH 7.2 (0.05% sodium azide).
- Immunogen:Recombinant protein corresponding to amino acids 792-1144 of Zika virus NS1.
- Purification:DEAE column chromatography
- Size:500 µG
- Pk:500 µG
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant Zika virus NS1.
Type: Primary
Antigen: NS1
Clonality: Monoclonal
Clone: M86641
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Viruses