Order Entry
Spain
ContactUsLinkComponent
Puller/inserter tool, CarboFrit®
Puller/inserter tool, CarboFrit®
Catalog # REST21642
Supplier:  Restek
undefined
Puller/inserter tool, CarboFrit®
Catalog # REST21642
Supplier:  Restek
Specifications
  • Description:
    CarboFrit puller/inserter tool
  • Pk:
    1
Specifications
About this item
This puller/inserter tool has hook end for removing CarboFrit® inserts and a bent end (90°) for inserting CarboFrit® inserts.
  • Slender design
  • Easy to maneuver