Order Entry
Spain
ContactUsLinkComponent
HIV-1 gp120 MN (from Baculovirus (insect cells))
HIV-1 gp120 MN (from Baculovirus (insect cells))
Supplier:  Biorbyt
Certificates
HIV-1 gp120 MN (from Baculovirus (insect cells))
Supplier:  Biorbyt

Specifications

  • Source:
    Baculovirus (Insect cells)
  • Storage conditions:
    Recombinant HIV -1 gp120 MN although stable at 4 °C for weeks, should be stored desiccated below −18 °C. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA). Please avoid freeze-thaw cycles.
  • Protein/peptide name:
    HIV-1 gp120 MN
  • Purity:
    Greater than 90.0% as determined by HPLC analysis and SDS-PAGE.
  • Formulation:
    The HIV-1 gp120 protein solution contains 30mM Tris, pH-7.6, 150mM NaCl and 0.01% Triton N101.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together