Rat recombinant CNTF (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Rat
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Protein/peptide name:CNTF
- Purity:> 95% by SDS-PAGE and HPLC analyses
- Molecular weight:22.7 kDa
- Sequence:AFAEQTPLTL HRRDLCSRSI WLARKIRSDL TALMESYVKH QGLNKNINLDSVDGVPVAST DRWSEMTEAE RLQENLQAYR TFQGMLTKLL EDQRVHFTPTEGDFHQAIHT LMLQVSAFAY QLEELMVLLE QKIPENEADG MPATVGDGGLFEKKLWGLKV LQELSQWTVR SIHDLRVISS HQMGISALES HYGAKDKQM
- Formulation:Lyophilized from a 0.2 um filtered concentrated solution in 2xPBS, pH 7.4, 2mM C-mercaptoethanol.
- Tested applications:SDS-PAGE , HPLC
Specifications
Frequently Bought Together