Order Entry
Spain
ContactUsLinkComponent
Rat recombinant FGF18 (from E. coli)
Rat recombinant FGF18 (from E. coli)
Supplier:  Biorbyt
Certificates
Rat recombinant FGF18 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Protein synonyms:
    Fibroblast Growth Factor-18
  • Protein/peptide name:
    FGF18
  • Purity:
    > 95% by SDS-PAGE and HPLC analyses
  • Molecular weight:
    21.0 kDa
  • Sequence:
    EENVDFRIHV ENQTRARDDV SRKQLRLYQL YSRTSGKHIQ VLGRRISARG EDGDKYAQLLVETDTFGSQV RIKGKETEFY LCMNRKGKLV GKPDGTSKEC VFIEKVLENN YTALMSAKYSGWYVGFTKKG RPRKGPKTRE NQQDVHFMKR YPKGQTELQK PFKYTTVTKRSRRIRPTHPG
  • Formulation:
    Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4, 500mM NaCl
  • Tested applications:
    SDS-PAGE , HPLC

Specifications

Related Information

Frequently Bought Together