Order Entry
Spain
ContactUsLinkComponent
 
Rhesus macaque recombinant IL3 (from E. coli)
 :  Biorbyt
 
Rhesus macaque recombinant IL3 (from E. coli)
 :  Biorbyt

 

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rhesus monkey
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Protein synonyms:
    Rhesus Macaque Interleukin-3
  • Protein/peptide name:
    IL3
  • Purity:
    > 98 % by SDS-PAGE and HPLC analyses
  • Molecular weight:
    14.0 kDa
  • Sequence:
    APMTQTTSLK TSWAKCSNMI DEIITHLNQP PLPSPDFNNL NEEDQTILVE KNLRRSNLEAFSKAVKSLQN ASAIESILKN LPPCLPMATA APTRPPIRIT NGDRNDFRRK LKFYLKTLENEQAQ
  • Formulation:
    Lyophilized from a 0.2 m filtered concentrated solution in PBS, pH 7.4, 5 % trehalose.
  • Tested applications:
    SDS-PAGE , HPLC

 

 

Frequently Bought Together