Order Entry
Spain
ContactUsLinkComponent
Recombinant TPO (from cells)
Recombinant TPO (from cells)
Catalog # BIRBORB79693-0.1
Supplier:  Biorbyt
CAS Number:  
Recombinant TPO (from cells)
Catalog # BIRBORB79693-0.1
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    0,1 mg
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Storage conditions:
    Store at −20 °C. in lyophilized form. After reconstitution the solution should be stored at −70 °C.Avoid repeated freezing and thawing.It is recommended to aliquot the product after reconstitution.
  • Protein synonyms:
    MSA
  • Protein/peptide name:
    TPO
  • Purity:
    Greater than 95% pure SDS-PAGE. Affinity and ion exchange chromatography
  • Formulation:
    Lyophilized from 20 mM Tris-HCl, pH 8.0, 250 mM NaCl, 0.1 mM KI, 5% D-mannitol. Product is recommended to reconstitute with deionized water.

Specifications

About this item