Order Entry
Spain
ContactUsLinkComponent
Human recombinant UBE2S (from E. coli)
Human recombinant UBE2S (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant UBE2S (from E. coli)
Supplier:  Biorbyt

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    UbiquitinConjugating Enzyme E2 S
  • UniProtKB:
    Q16763 (Human)
  • Protein/peptide name:
    UBE2S
  • Purity:
    >95% as determined by reducing SDS-PAGE.
  • Sequence:
    MSPILGYWKIKGLVQPTRLLLEYLEEKYEEHLYERDEGDKWRNKKFELGLEFPNLPYYIDGDVKL TQSMAIIRYIADKHNMLGGCPKERAEISMLEGAVLDIRYGVSRIAYSKDFETLKVDFLSKLPEML KMFEDRLCHKTYLNGDHVTHPDFMLYDALDVVLYMDPMCLDAFPKLVCFKKRIEAIPQIDKYLKS SKYIAWPLQGWQATFGGGDHPPKSDLVPRGSMNSNVENLPPHIIRLVYKEVTTLTADPPDGIKVF PNEEDLTDLQVTIEGPEGTPYAGGLFRMKLLLGKDFPASPPKGYFLTKIFHPNVGANGEICVNVL KRDWTAELGIRHVLLTIKCLLIHPNPESALNEEAGRLLLENYEEYAARARLLTEIHGGAGGPSGR AEAGRALASGTEASSTDPGAPGGPGGAEGPMAKKHAGERDKKLAAKKKTDKKRALRRL
  • Formulation:
    Supplied as a 0.2 um filtered solution of 50mM HEPES, 150mM NaCl, 2mM DTT, 10% Glycerol, pH 7.5.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together